Learn More
Medchemexpress LLC 7-Ethoxyresorufin | 5725-91-7 | 99.6% | 50 MG
SDP
Catalog No. 5000409678
Encompass_Preferred
7-Ethoxyresorufin, also known as Resorufin ethyl ether, is a fluorometric substrate and competitive inhibitor of cytochrome P450, specifically CYP1A1. It also inhibits NO synthase. This compound has been shown to reduce the relaxant actions of acetylcholine in endothelium-intact rings and of NO, GTN, and SNP in endothelium-denuded rings in rat aortic rings.
  • Acts as a fluorometric substrate
  • Functions as a competitive inhibitor of cytochrome P450, particularly CYP1A1
  • Inhibits NO synthase
  • Reduces the relaxant actions of acetylcholine, NO, GTN, and SNP
  • Reacts with NADPH to produce a fluorescent product
Catalog No. 50-004-09678 Supplier Medchemexpress LLC Supplier No. HYD014550MG
7-Ethoxyresorufin, also known as Resorufin ethyl ether, is a fluorometric substrate and competitive inhibitor of cytochrome P450, specifically CYP1A1. It also inhibits NO synthase. This compound has been shown to reduce the relaxant actions of acetylcholine in endothelium-intact rings and of NO, GTN, and SNP in endothelium-denuded rings in rat aortic rings.
  • Acts as a fluorometric substrate
  • Functions as a competitive inhibitor of cytochrome P450, particularly CYP1A1
  • Inhibits NO synthase
  • Reduces the relaxant actions of acetylcholine, NO, GTN, and SNP
  • Reacts with NADPH to produce a fluorescent product