Learn More
Medchemexpress LLC 7-Ethoxyresorufin (resorufin ethyl ether) | 5725-91-7 | 99.6% | 1 MG
SDP
Catalog No. 5000410845
Encompass_Preferred
7-Ethoxyresorufin (Resorufin ethyl ether) is a fluorometric substrate and competitive inhibitor of cytochrome P450, particularly CYP1A1. It also inhibits NO synthase.
  • Reduces the relaxant actions of acetylcholine in endothelium-intact rings and of NO, GTN, and SNP in endothelium-denuded rings in a concentration-dependent manner.
  • Reacts with NADPH in the presence of acid to form a product that fluoresces at the same wavelength as Resorufin.
Catalog No. 50-004-10845 Supplier Medchemexpress LLC Supplier No. HYD01451MG
7-Ethoxyresorufin (Resorufin ethyl ether) is a fluorometric substrate and competitive inhibitor of cytochrome P450, particularly CYP1A1. It also inhibits NO synthase.
  • Reduces the relaxant actions of acetylcholine in endothelium-intact rings and of NO, GTN, and SNP in endothelium-denuded rings in a concentration-dependent manner.
  • Reacts with NADPH in the presence of acid to form a product that fluoresces at the same wavelength as Resorufin.