Learn More
Medchemexpress LLC 7-Ethoxyresorufin (Resorufin ethyl ether) | 5725-91-7 | 99.6% | 5 MG
SDP
Catalog No. 5000411050
Encompass_Preferred
7-Ethoxyresorufin (Resorufin ethyl ether) is a fluorometric substrate and competitive inhibitor of cytochrome P450, specifically CYP1A1. It also acts as an inhibitor of NO synthase. The product is intended for research use only.
  • Fluorometric substrate
  • Competitive inhibitor of cytochrome P450, especially CYP1A1
  • Inhibits NO synthase
Catalog No. 50-004-11050 Supplier Medchemexpress LLC Supplier No. HYD01455MG
7-Ethoxyresorufin (Resorufin ethyl ether) is a fluorometric substrate and competitive inhibitor of cytochrome P450, specifically CYP1A1. It also acts as an inhibitor of NO synthase. The product is intended for research use only.
  • Fluorometric substrate
  • Competitive inhibitor of cytochrome P450, especially CYP1A1
  • Inhibits NO synthase