Learn More
Medchemexpress LLC 7-ethyl-4-[3-(4-ethylsulfonyl-2-methoxyphenyl)-4-fluorophenyl]imidazo[4,5-c]pyridazine | 1614245-70-3 | 99.3% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY120874100MG
Encompass_Preferred
Darigabat is an orally active, subtype-selective GABAA positive allosteric modulator (PAM) used in preclinical and research studies to investigate GABAA receptor pharmacology, anxiolytic effects, and anticonvulsant potential.
  • Subtype-selective GABAA PAM targeting α2/α3/α5 subunits.
  • High purity (99.3%) suitable for research applications.
  • Blood-brain barrier penetrant with demonstrated anxiolytic activity in models.
  • Available as solid or DMSO solution; multiple package sizes offered.
  • Useful for pharmacology, pain, anxiety, and epilepsy research.
Catalog No. 50-002-30822