Learn More
Medchemexpress LLC 7-ethyl-4-[3-(4-ethylsulfonyl-2-methoxyphenyl)-4-fluorophenyl]imidazo[4,5-c]pyridazine | 1614245-70-3 | 99.3% | 25 MG
SDP
Catalog No. 5000233883
Encompass_Preferred
Darigabat is an orally active, subtype-selective GABAA positive allosteric modulator used in neuroscience and pharmacology research. It has molecular formula C22H21FN4O3S and molecular weight 440.49 g/mol, and is supplied in small research quantities with high purity.
  • Orally active GABAA positive allosteric modulator.
  • Subtype-selective for α2/α3/α5 receptor subunits.
  • High purity of 99.3% suitable for research applications.
  • Molecular weight 440.49 g/mol and formula C22H21FN4O3S.
  • Supplied in small-quantity packages for in vitro and in vivo studies.
Catalog No. 50-002-33883 Supplier Medchemexpress LLC Supplier No. HY12087425MG
Darigabat is an orally active, subtype-selective GABAA positive allosteric modulator used in neuroscience and pharmacology research. It has molecular formula C22H21FN4O3S and molecular weight 440.49 g/mol, and is supplied in small research quantities with high purity.
  • Orally active GABAA positive allosteric modulator.
  • Subtype-selective for α2/α3/α5 receptor subunits.
  • High purity of 99.3% suitable for research applications.
  • Molecular weight 440.49 g/mol and formula C22H21FN4O3S.
  • Supplied in small-quantity packages for in vitro and in vivo studies.