Learn More
Medchemexpress LLC 7-Ethylcamptothecin | 78287-27-1 | 250 MG
SDP
Catalog No. 5000420690
Encompass_Preferred
7-Ethylcamptothecin is a derivative of Camptothecin and a DNA topoisomerase I inhibitor. It exhibits significant antitumor activity against tumor cells and serves as a key precursor for the synthesis of 20 (S)-O-substituted benzoyl-7-ethylcamptothecin compounds. It is widely used in various tumor-related scientific studies.
  • Functions as a DNA topoisomerase I inhibitor.
  • Exhibits significant antitumor activity against tumor cells.
  • Serves as a precursor for synthesizing 20 (S)-O-substituted benzoyl-7-ethylcamptothecin compounds.
  • Utilized in various tumor-related scientific studies.
  • Starting material for developing E-ring modified water-soluble camptothecin derivatives.
  • Appears as a light yellow to yellow solid.
  • Has a purity of 99.58%.
Catalog No. 50-004-20690 Supplier Medchemexpress LLC Supplier No. 250MGHYN2108
7-Ethylcamptothecin is a derivative of Camptothecin and a DNA topoisomerase I inhibitor. It exhibits significant antitumor activity against tumor cells and serves as a key precursor for the synthesis of 20 (S)-O-substituted benzoyl-7-ethylcamptothecin compounds. It is widely used in various tumor-related scientific studies.
  • Functions as a DNA topoisomerase I inhibitor.
  • Exhibits significant antitumor activity against tumor cells.
  • Serves as a precursor for synthesizing 20 (S)-O-substituted benzoyl-7-ethylcamptothecin compounds.
  • Utilized in various tumor-related scientific studies.
  • Starting material for developing E-ring modified water-soluble camptothecin derivatives.
  • Appears as a light yellow to yellow solid.
  • Has a purity of 99.58%.