Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Fluoro-2,3,4,9-tetrahydro-1H-beta-carboline, Thermo Scientific™

Catalog No. MAY00133DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAY00133DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAY00133DA Supplier Thermo Scientific Chemicals Supplier No. MAY00133DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 177858-80-9
Molecular Formula C11H11FN2
Molecular Weight (g/mol) 190.221
InChI Key OTGGOPSBNVEBDA-UHFFFAOYSA-N
Synonym 7-fluoro-2,3,4,9-tetrahydro-1h-beta-carboline, 7-fluoro-1,2,3,4-tetrahydro-9h-pyrido 3,4-b indole, 7-fluoro-1h,2h,3h,4h,9h-pyrido 3,4-b indole, 7-fluoro-2,3,4,9-tetrahydro-1h-pyrido 3,4-b indole, 7-fluoro-1,2,3,4-tetrahydrobeta-carboline, 1h-pyrido 3,4-b indole, 7-fluoro-2,3,4,9-tetrahydro
PubChem CID 15077550
IUPAC Name 7-fluoro-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole
SMILES C1CNCC2=C1C3=C(N2)C=C(C=C3)F

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 190.22
Percent Purity 95%
Chemical Name or Material 7-Fluoro-2,3,4,9-tetrahydro-1H-β-carboline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.