Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-Fluoro-2-tetralone | 29419-15-6 | 95.0% | 164.18 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY30051100MG

Encompass_Preferred

7-Fluoro-2-tetralone is a drug intermediate used for the synthesis of various active compounds. It is intended for research use only and is not sold to patients.

  • Molecular weight: 164.18
  • Formula: C10H9FO
  • Appearance: Solid below 58°C, liquid above 58.5°C
  • Color: Light yellow to brown
  • SMILES: O=C1CC(C=C(F)C=C2)=C2CC1
  • Shipping: Room temperature within the continental US; may vary elsewhere.
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years.
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month.
  • Purity: 95.0%

Catalog No. 50-004-04321


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.