Learn More
Medchemexpress LLC 7-fluoro-6-methoxy-4-[6-[4-(4-methylpiperazin-1-yl)piperidin-1-yl]pyrazolo[1,5-a]pyrimidin-3-yl]quinol | 2248154-85-8 | MFCD32859311 | 99.4% | C26H30FN7O | 10MG
SDP
Catalog No. 5000260000
Encompass_Preferred
KER047 (ALK2-IN-4) is a potent small-molecule inhibitor of activin receptor-like kinase-2 (ALK2) intended for research use in the study of ALK2-mediated signaling and metabolic disorders such as fibrodysplasia ossificans progressiva (FOP). The compound is provided in solid and solution formats for in vitro and preclinical investigations.
  • Potent ALK2 inhibitor with demonstrated activity in research assays.
  • Suitable for research into metabolic disease models, including FOP.
  • High purity (99.4%) for reproducible experimental results.
  • Available as solid powder and 10 mM solutions in DMSO for flexible dosing.
  • Chemical formula C26H30FN7O and molecular weight 475.56 g/mol.
  • Stable storage recommendations: powder at -20°C or 4°C; in solvent at -80°C or -20°C.
Catalog No. 50-002-60000 Supplier Medchemexpress LLC Supplier No. HY13677310MG
KER047 (ALK2-IN-4) is a potent small-molecule inhibitor of activin receptor-like kinase-2 (ALK2) intended for research use in the study of ALK2-mediated signaling and metabolic disorders such as fibrodysplasia ossificans progressiva (FOP). The compound is provided in solid and solution formats for in vitro and preclinical investigations.
  • Potent ALK2 inhibitor with demonstrated activity in research assays.
  • Suitable for research into metabolic disease models, including FOP.
  • High purity (99.4%) for reproducible experimental results.
  • Available as solid powder and 10 mM solutions in DMSO for flexible dosing.
  • Chemical formula C26H30FN7O and molecular weight 475.56 g/mol.
  • Stable storage recommendations: powder at -20°C or 4°C; in solvent at -80°C or -20°C.