Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Fluoro-6-nitro-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific Chemicals

Catalog No. AAH61065MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61065MD 250 mg
AAH6106503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH61065MD Supplier Thermo Scientific Chemicals Supplier No. H61065MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 122138-83-4 | Molecular Formula: C8H7FN2O3 | Molecular Weight: 198.153 g/mol

This compounds are useful laboratory reagents and used frequently in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This compounds are useful laboratory reagents and used frequently in organic synthesis.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122138-83-4
Molecular Formula C8H7FN2O3
Molecular Weight (g/mol) 198.153
MDL Number MFCD23379858
InChI Key PEOVRVSERLAIQM-UHFFFAOYSA-N
PubChem CID 14422956
IUPAC Name 7-fluoro-6-nitro-3,4-dihydro-2H-1,4-benzoxazine
SMILES C1COC2=CC(=C(C=C2N1)[N+](=O)[O-])F

Specifications

Quantity 250 mg
Formula Weight 198.15
Percent Purity 97%
Chemical Name or Material 7-Fluoro-6-nitro-3,4-dihydro-2H-1,4-benzoxazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.