Learn More
Medchemexpress LLC 7-fluorobenzofurazan-4-sulfonic acid ammonium | 84806-27-9 | 99.0% | 235.19 | C6H6FN3O4S | 10 MG
SDP
Catalog No. 5000297315
Encompass_Preferred
7-Fluorobenzofurazan-4-sulfonic acid ammonium is a fluorescent labeling reagent (ammonium salt) used for derivatization and sensitive detection of thiol-containing compounds in analytical assays.
  • Fluorescent reagent for detecting thiol-disulfide species such as cysteine and glutathione.
  • High reported purity (~98.9%), suitable for analytical use.
  • Available in small research-scale quantities for laboratory experiments.
  • Compatible with pre-column derivatization and HPLC fluorescence detection methods.
  • For research use only; not intended for human use.
Catalog No. 50-002-97315 Supplier Medchemexpress LLC Supplier No. HYD009710MG
7-Fluorobenzofurazan-4-sulfonic acid ammonium is a fluorescent labeling reagent (ammonium salt) used for derivatization and sensitive detection of thiol-containing compounds in analytical assays.
  • Fluorescent reagent for detecting thiol-disulfide species such as cysteine and glutathione.
  • High reported purity (~98.9%), suitable for analytical use.
  • Available in small research-scale quantities for laboratory experiments.
  • Compatible with pre-column derivatization and HPLC fluorescence detection methods.
  • For research use only; not intended for human use.