Learn More
Medchemexpress LLC 7-Fluorobenzofurazan-4-sulfonic acid (ammonium) | 84806-27-9 | 99.0% | 25 MG
SDP
Catalog No. 5000413555
Encompass_Preferred
7-Fluorobenzofurazan-4-sulfonic acid ammonium is a fluorescent label primarily used for detecting dissolved thiol-disulfide compounds such as Cys and GSH. It is suitable for various laboratory applications requiring fluorescence-based detection.
  • Fluorescent label for thiol-disulfide detection
  • Used with Cys and GSH
  • Research use only
  • Excitation wavelength range: 451-495 nm Blue
  • Emission wavelength range: 451-495 nm Blue
Catalog No. 50-004-13555 Supplier Medchemexpress LLC Supplier No. 25MGHYD0097
7-Fluorobenzofurazan-4-sulfonic acid ammonium is a fluorescent label primarily used for detecting dissolved thiol-disulfide compounds such as Cys and GSH. It is suitable for various laboratory applications requiring fluorescence-based detection.
  • Fluorescent label for thiol-disulfide detection
  • Used with Cys and GSH
  • Research use only
  • Excitation wavelength range: 451-495 nm Blue
  • Emission wavelength range: 451-495 nm Blue