Learn More
US Plastic Corporation White 7 Gallon Bucket
SDP
Catalog No. NC9836505
Encompass
These round pails are tough enough to store nails and secure enough to protect contents. HDPE bucket. Strong reinforcing ribs. Space-saving, tapered design allows pails to nest. Wall thickness: 0.090" ±0.005”. Dimensions: 11.625" Top OD X 19.375" Hgt. Can be hot filled up to 190°F & can also be frozen. Meets FDA standards. Buckets & lids sold separately
Catalog No. NC9836505 Supplier US Plastic Corporation Supplier No. 2393
These round pails are tough enough to store nails and secure enough to protect contents. HDPE bucket. Strong reinforcing ribs. Space-saving, tapered design allows pails to nest. Wall thickness: 0.090" ±0.005”. Dimensions: 11.625" Top OD X 19.375" Hgt. Can be hot filled up to 190°F & can also be frozen. Meets FDA standards. Buckets & lids sold separately