Learn More
Enzo Life Sciences 7-HC-6-heptenoate (10 mg)
SDP
Supplier:  Enzo Life Sciences BMLST5010010
Encompass_Preferred
PLA2 fluorogenic substrate for use in assays without phospholipid and Ca2+. For use with purified enzyme, as this substrate is nonspecific. Ideal for drug discovery, HTS, and similar applications. Ex.: 360 nm, Em.: 460 nm Purity: ≥98% (TLC). Solubility: Soluble in DMSO (50 mg/ml) or ethanol (35 mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-2186