Learn More
Enzo Life Sciences 7-HC-arachidonate (10 mg). CAS: 161180-11-6
SDP
Catalog No. 502012187
Encompass_Preferred
PLA2 fluorogenic substrate for use in assays with phospholipid and Ca2+. For use with purified enzyme, as this substrate is nonspecific. Ideal for drug discovery, HTS, and similar applications. Ex.: 360 nm, Em.: 460 nm Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (50mg/ml) or hexane. Use only de-gassed exane.   Do not use ethanol as it may degrade product. Long Term Storage: -20°C.
Catalog No. 50-201-2187 Supplier Enzo Life Sciences Supplier No. BMLST5020010
PLA2 fluorogenic substrate for use in assays with phospholipid and Ca2+. For use with purified enzyme, as this substrate is nonspecific. Ideal for drug discovery, HTS, and similar applications. Ex.: 360 nm, Em.: 460 nm Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (50mg/ml) or hexane. Use only de-gassed exane.   Do not use ethanol as it may degrade product. Long Term Storage: -20°C.