Learn More
AdipoGen 7-Hydroxy-4-coumarinylacetic acid (5 g)
SDP
Supplier:  AdipoGen CDXH0008G005
Encompass_Preferred
Chemical. CAS: 6950-82-9. Formula: C11H8O5. MW: 220.18. 7-Hydroxycoumarin-4-acetic acid is a fluorescent pH indicator. 7-Hydroxycoumarin-4-acetic acid is used for labeling amine-specific peptides, proteins and other biological molecules to give blue fluorescence. It is used as fluorigenic substrate, e.g. in BSA-HCA complexes for fatty acid concentration measurement or for fluorometric enzymatic analysis. It also can be used as a building block for synthesis of other coumarin-based probes. Spectral data: lambdaEx=360nm; lambdaEm=450nm (DMSO).
Catalog No. 50-205-4485