Learn More
Electron Microscopy Sciences Embra 7 Hexagonal Grid 7-Hex, Copper
SDP
Catalog No. 5024372
Encompass_Preferred
Embra electroformed grids are 3.05 mm in diameter and 16 um thick.
Type: 7-Hex
Material: Copper
Catalog No. 50-243-72 Supplier Electron Microscopy Sciences Supplier No. E7HEXCU
Embra electroformed grids are 3.05 mm in diameter and 16 um thick.
Type: 7-Hex
Material: Copper