Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-hydrazinyl-4-methyl-2H-chromen-2-one | 113707-87-2 | MFCD09999551 | 97.1% | 190.20 | C10H10N2O2 | 100MG
SDP

Catalog No. 5000238447
Encompass_Preferred

Coumarin hydrazine is a fluorescent chemical probe used to label protein- and lipid-bound carbonyls in cells and biochemical assays. It reacts with aldehydes and ketones to form fluorescent hydrazones, producing a measurable signal suitable for live-cell and in vitro labeling when handled protected from light (λex = 420-450 nm; λem ≈ 468 nm).

  • Fluorogenic probe for labeling protein- and lipid-bound carbonyls.
  • Reacts with aldehydes and ketones to form fluorescent hydrazones.
  • Excitation 420-450 nm; emission ≈468 nm.
  • Molecular formula C10H10N2O2; molecular weight 190.20.
  • Purity 97.1%; supplied as a 100 mg solid.
  • Protect from light; in solvent store at -80°C for up to 6 months or at -20°C for up to 1 month.

Catalog No. 50-002-38447 Supplier Medchemexpress LLC Supplier No. HY124197100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Coumarin hydrazine is a fluorescent chemical probe used to label protein- and lipid-bound carbonyls in cells and biochemical assays. It reacts with aldehydes and ketones to form fluorescent hydrazones, producing a measurable signal suitable for live-cell and in vitro labeling when handled protected from light (λex = 420-450 nm; λem ≈ 468 nm).

  • Fluorogenic probe for labeling protein- and lipid-bound carbonyls.
  • Reacts with aldehydes and ketones to form fluorescent hydrazones.
  • Excitation 420-450 nm; emission ≈468 nm.
  • Molecular formula C10H10N2O2; molecular weight 190.20.
  • Purity 97.1%; supplied as a 100 mg solid.
  • Protect from light; in solvent store at -80°C for up to 6 months or at -20°C for up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.