Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-hydroxy-2,3,4,5-tetrahydro-benzofuro[2,3-c]azepin-1-one | 521937-07-5 | 99.1% | 100 MG
SDP

Catalog No. 5000233690
Encompass_Preferred

This small-molecule inhibitor targets protein kinase D (PKD) isoforms with submicromolar potency and is supplied as a white to off-white solid for research use.

  • Inhibits PKD1, PKD2, and PKD3 with IC50s of 182 nM, 280 nM, and 227 nM.
  • Purity 99.1% (HPLC).
  • Molecular formula C12H11NO3; molecular weight 217.22 g/mol.
  • Soluble in DMSO at 100 mg/mL; in vivo formulations achievable at ≥2.5 mg/mL using recommended vehicles.
  • Appearance white to off-white solid.
  • Storage: powder -20°C up to 3 years; in solvent -80°C up to 2 years.
  • Supplied as a 100 mg pack; also available in smaller and larger sizes or as 10 mM DMSO solutions.

Catalog No. 50-002-33690 Supplier Medchemexpress LLC Supplier No. HY12239100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This small-molecule inhibitor targets protein kinase D (PKD) isoforms with submicromolar potency and is supplied as a white to off-white solid for research use.

  • Inhibits PKD1, PKD2, and PKD3 with IC50s of 182 nM, 280 nM, and 227 nM.
  • Purity 99.1% (HPLC).
  • Molecular formula C12H11NO3; molecular weight 217.22 g/mol.
  • Soluble in DMSO at 100 mg/mL; in vivo formulations achievable at ≥2.5 mg/mL using recommended vehicles.
  • Appearance white to off-white solid.
  • Storage: powder -20°C up to 3 years; in solvent -80°C up to 2 years.
  • Supplied as a 100 mg pack; also available in smaller and larger sizes or as 10 mM DMSO solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.