Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 7-Hydroxy-3-(4-methoxyphenoxy)-4H-chromen-4-one

Catalog No. OTV0002731G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV0002731G 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV0002731G Supplier Sigma Aldrich Supplier No. OTV0002731G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Spearmint oil is extracted from aerial parts of mentha spicata L
  • Spearmint oil has antimicrobial property
  • This essential oil is used in toothpaste as a flavoring ingredient
  • In addition, spearmint oil is also used in chewing gum and medicines.download our flavors and fragrances catalog to view our entire product line.subscribe to our newsletter to keep up to date on our latest flavors and fragrances offerings

Specifications

CAS 137987-99-6
Molecular Formula C16H12O5
Linear Formula C16H12O5
MDL Number MFCD00525549
Quantity 10 mL
Molecular Weight (g/mol) 284.26
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.