Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Hydroxy-3,7-dimethyloctanal, MP Biomedicals

Catalog No. ICN224939 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN224939 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN224939 Supplier MP Biomedicals, Inc Supplier No. 0522493910
Add to Cart
Add to Cart

Chemical Identifiers

CAS 107-75-5
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.268
InChI Key WPFVBOQKRVRMJB-UHFFFAOYSA-N
Synonym hydroxycitronellal, 7-hydroxycitronellal, citronellal hydrate, 3,7-dimethyl-7-hydroxyoctanal, phixia, cyclalia, cyclosia, laurine, fixol, lilyl aldehyde
PubChem CID 7888
ChEBI CHEBI:53459
IUPAC Name 7-hydroxy-3,7-dimethyloctanal
SMILES CC(CCCC(C)(C)O)CC=O

Specifications

Quantity 10 g
Formula Weight 172
Chemical Name or Material 7-Hydroxy-3, 7-Dimethyl Octanal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.