Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Hydroxy-4-cholesten-3-one, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014101 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4101 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4101 Supplier MilliporeSigma Supelco Supplier No. 528531MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7 α-Hydroxy-4-cholesten-3-one (C4) is known as a marker for the cholesterol 7 α-hydroxylase activity. It plays an important role in bile acid metabolism.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3862-25-7
Linear Formula C27H44O2
MDL Number MFCD00870345
Synonym 7 alpha-Hydroxycholest-4-en-3-one
Type Analytical Standard
Quantity 1 mg
Formula Weight 400.64
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.