Learn More
Aat Bioquest 7-Hydroxy-4-methylcoumarin [4-Methylumbelliferone] *Fluorescence reference standard*
SDP
Catalog No. 501952679
Encompass_Preferred
This compound is used as a calibration standard for 7-hydroxy-4-methylcoumarin-based enzyme substrates. It has pH-dependent fluorescence.
Catalog No. 50-195-2679 Supplier Aat Bioquest Supplier No. 41
This compound is used as a calibration standard for 7-hydroxy-4-methylcoumarin-based enzyme substrates. It has pH-dependent fluorescence.