Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Hydroxy-4'-nitroisoflavone, 98%, Thermo Scientific™

Catalog No. AAH6416706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6416706 5 g
AAH6416714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6416706 Supplier Thermo Scientific Chemicals Supplier No. H6416706

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
7-Hydroxy-4′-nitroisoflavone is used in the synthesis of iso flavones, Synthesis of daidzin, soflavone amide derivatives.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15485-80-0
Molecular Formula C15H9NO5
Molecular Weight (g/mol) 283.24
MDL Number MFCD01546432
InChI Key CEESSYSSRMOMDE-UHFFFAOYSA-N
Synonym 7-hydroxy-4'-nitroisoflavone, 7-hydroxy-3-4-nitrophenyl-4h-chromen-4-one, 7-hydroxy-3-4-nitrophenyl chromen-4-one, 7-hydroxy-3-4-nitrophenyl-4h-1-benzopyran-4-one, 7-hydroxy-4-nitroisoflavone, 7-hydroxy-4/'-nitroisoflavone, 7-hydroxy-4'-nitro-isoflavone 3c
PubChem CID 7454026
IUPAC Name 7-hydroxy-3-(4-nitrophenyl)chromen-4-one
SMILES OC1=CC=C2C(=O)C(=COC2=C1)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 290°C to 292°C
Color Yellow
Quantity 5 g
Beilstein 385006
Solubility Information Slightly soluble in water.
Formula Weight 283.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 7-Hydroxy-4'-nitroisoflavone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.