Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-hydroxy-4-phenylcoumarin | 2555-30-8 | MFCD00024080 | 96.9% | 238.24 g/mol | C15H10O3 | 1 ML
SDP

Catalog No. 5000280631
Encompass_Preferred

7-Hydroxy-4-phenylcoumarin (4-phenylumbelliferone) is a small-molecule research reagent reported to inhibit aldehyde dehydrogenase 2 (ALDH-2) and monoamine oxidase (MAO). It is supplied as a solution in DMSO and as a solid for use in biochemical assays and enzyme inhibition studies; handle according to the SDS and laboratory safety practices.

  • Purity: 96.89%.
  • Molecular weight: 238.24 g/mol.
  • Chemical formula: C15H10O3.
  • CAS number: 2555-30-8.
  • Available as a 10 mM solution in DMSO (1 mL) and as a solid.
  • Biological activity: dual ALDH-2 and MAO inhibitor (IC50 1.5 μM and 0.5 μM).
  • Intended use: research reagent for biochemical assays and enzyme inhibition studies.

Catalog No. 50-002-80631 Supplier Medchemexpress LLC Supplier No. HYW20719510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Hydroxy-4-phenylcoumarin (4-phenylumbelliferone) is a small-molecule research reagent reported to inhibit aldehyde dehydrogenase 2 (ALDH-2) and monoamine oxidase (MAO). It is supplied as a solution in DMSO and as a solid for use in biochemical assays and enzyme inhibition studies; handle according to the SDS and laboratory safety practices.

  • Purity: 96.89%.
  • Molecular weight: 238.24 g/mol.
  • Chemical formula: C15H10O3.
  • CAS number: 2555-30-8.
  • Available as a 10 mM solution in DMSO (1 mL) and as a solid.
  • Biological activity: dual ALDH-2 and MAO inhibitor (IC50 1.5 μM and 0.5 μM).
  • Intended use: research reagent for biochemical assays and enzyme inhibition studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.