Learn More
Medchemexpress LLC HY-N6596 50mg Medchemexpress, 7-Hydroxy-4H-chromen-4-one CAS:59887-89-7 Purity:>98%
SDP
Catalog No. 501933960
Encompass_Preferred
Medchemexpress, HY-N6596 50mg 7-Hydroxy-4H-chromen-4-one CAS:59887-89-7 7-Hydroxychromone is a Src kinase inhibitor with an IC 50 of <300 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3960 Supplier Medchemexpress LLC Supplier No. HYN659650MG
Medchemexpress, HY-N6596 50mg 7-Hydroxy-4H-chromen-4-one CAS:59887-89-7 7-Hydroxychromone is a Src kinase inhibitor with an IC 50 of <300 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.