Learn More
Cayman Chemical 7-hydroxy Methotrexate 10mg
SDP
Catalog No. 502948388
Encompass_Preferred
A metabolite of methotrexate; does not inhibit thymidylate synthase in isolated human acute myeloid and acute lymphoblastic leukemia cells; has a maximum tolerated dose of 0.1 g/kg in rats
Catalog No. 50-294-8388 Supplier Cayman Chemical Supplier No. 3323910 MG
A metabolite of methotrexate; does not inhibit thymidylate synthase in isolated human acute myeloid and acute lymphoblastic leukemia cells; has a maximum tolerated dose of 0.1 g/kg in rats