Learn More
Cayman Chemical 7-hydroxy Warfarin, Purity: ≥98%, CAS: 17834-03-6, 10 mg
SDP
Catalog No. 502894734
Encompass_Preferred
A phase I metabolite of (+)-warfarin; a product of CYP2C9 metabolism of (-)-warfarin
Catalog No. 50-289-4734 Supplier Cayman Chemical Supplier No. 900067210 MG
A phase I metabolite of (+)-warfarin; a product of CYP2C9 metabolism of (-)-warfarin