Learn More
Cayman Chemical 7-hydroxy WarfarIn 1mg
SDP
Catalog No. 502893922
Encompass_Preferred
A phase I metabolite of (+)-warfarin; a product of CYP2C9 metabolism of (-)-warfarin
Catalog No. 50-289-3922 Supplier Cayman Chemical Supplier No. 90006721 MG
A phase I metabolite of (+)-warfarin; a product of CYP2C9 metabolism of (-)-warfarin