Learn More
Cayman Chemical 7-hydroxy WarfarIn 500UG
SDP
Catalog No. 502894922
Encompass_Preferred
A phase I metabolite of (+)-warfarin; a product of CYP2C9 metabolism of (-)-warfarin
Catalog No. 50-289-4922 Supplier Cayman Chemical Supplier No. 9000672500 UG
A phase I metabolite of (+)-warfarin; a product of CYP2C9 metabolism of (-)-warfarin