Learn More
Cayman Chemical 7-hydroxy WarfarIn 5mg
SDP
Catalog No. 502894777
Encompass_Preferred
A phase I metabolite of (+)-warfarin; a product of CYP2C9 metabolism of (-)-warfarin
Catalog No. 50-289-4777 Supplier Cayman Chemical Supplier No. 90006725 MG
A phase I metabolite of (+)-warfarin; a product of CYP2C9 metabolism of (-)-warfarin