Learn More
Medchemexpress LLC 7α-Hydroxycholesterol | 566-26-7 | 10 MG
SDP
Catalog No. 5000417296
Encompass_Preferred
7α-Hydroxycholesterol is a cholesterol oxide that can serve as a biomarker for oxidative stress and lipid peroxidation. It exhibits cytotoxic and pro-inflammatory activities. This compound can also inhibit sterol synthesis and reduce the activity of HMG-CoA reductase. It is used in the research of diseases such as diabetes and atherosclerosis.
  • Functions as a biomarker for oxidative stress and lipid peroxidation.
  • Possesses cytotoxic and pro-inflammatory activities.
  • Inhibits sterol synthesis.
  • Reduces the activity of HMG-CoA reductase.
  • Applicable in the research of diseases like diabetes and atherosclerosis.
Catalog No. 50-004-17296 Supplier Medchemexpress LLC Supplier No. HYN726410MG
7α-Hydroxycholesterol is a cholesterol oxide that can serve as a biomarker for oxidative stress and lipid peroxidation. It exhibits cytotoxic and pro-inflammatory activities. This compound can also inhibit sterol synthesis and reduce the activity of HMG-CoA reductase. It is used in the research of diseases such as diabetes and atherosclerosis.
  • Functions as a biomarker for oxidative stress and lipid peroxidation.
  • Possesses cytotoxic and pro-inflammatory activities.
  • Inhibits sterol synthesis.
  • Reduces the activity of HMG-CoA reductase.
  • Applicable in the research of diseases like diabetes and atherosclerosis.