Learn More
Medchemexpress LLC 7α-Hydroxycholesterol | 566-26-7 | 5 MG
SDP
Catalog No. 5000419189
Encompass_Preferred
7α-Hydroxycholesterol is a cholesterol oxide that serves as a biomarker for oxidative stress and lipid peroxidation. It exhibits cytotoxic and pro-inflammatory activities, can inhibit sterol synthesis, and reduce HMG-CoA reductase activity. This compound is applicable for research in diseases such as diabetes and atherosclerosis.
  • Serves as a biomarker for oxidative stress and lipid peroxidation
  • Exhibits cytotoxic and pro-inflammatory activities
  • Can inhibit sterol synthesis
  • Reduces HMG-CoA reductase activity
  • Applicable for research in diseases such as diabetes and atherosclerosis
Catalog No. 50-004-19189 Supplier Medchemexpress LLC Supplier No. HYN72645MG
7α-Hydroxycholesterol is a cholesterol oxide that serves as a biomarker for oxidative stress and lipid peroxidation. It exhibits cytotoxic and pro-inflammatory activities, can inhibit sterol synthesis, and reduce HMG-CoA reductase activity. This compound is applicable for research in diseases such as diabetes and atherosclerosis.
  • Serves as a biomarker for oxidative stress and lipid peroxidation
  • Exhibits cytotoxic and pro-inflammatory activities
  • Can inhibit sterol synthesis
  • Reduces HMG-CoA reductase activity
  • Applicable for research in diseases such as diabetes and atherosclerosis