Learn More
Indofine Chemical 7-Hydroxycoumarin, 99%, 250 Mg
SDP
Catalog No. 50908310
Encompass_Preferred
Molecular Formula: C10H6O5, Purity >99%, Molecular Weight: 206.16, Melting Point >270C, Appearance: Cream colored powder, Solubility: Soluble in DMF, Storage conditions: Store in a cool, dry place, Also known as 3-Carboxy-7-hydroxycoumarin
Catalog No. 50-908-310 Supplier Indofine Chemical Supplier No. 19146 250 MG
Molecular Formula: C10H6O5, Purity >99%, Molecular Weight: 206.16, Melting Point >270C, Appearance: Cream colored powder, Solubility: Soluble in DMF, Storage conditions: Store in a cool, dry place, Also known as 3-Carboxy-7-hydroxycoumarin