Learn More
Medchemexpress LLC 7-hydroxycoumarin-3-carboxylic acid N-succinimidyl ester | 134471-24-2 | 100 MG
SDP
Catalog No. 5000410049
Encompass_Preferred
7-Hydroxycoumarin-3-carboxylic acid N-succinimidyl ester is the amine-reactive succinimidyl ester of 7-Hydroxycoumarin-3-carboxylic acid. It functions as a blue fluorescent dye for labeling proteins and nucleic acids.
  • Amine-reactive succinimidyl ester
  • Blue fluorescent dye
  • Used for labeling proteins
  • Used for labeling nucleic acids
Catalog No. 50-004-10049 Supplier Medchemexpress LLC Supplier No. HYD0115100MG
7-Hydroxycoumarin-3-carboxylic acid N-succinimidyl ester is the amine-reactive succinimidyl ester of 7-Hydroxycoumarin-3-carboxylic acid. It functions as a blue fluorescent dye for labeling proteins and nucleic acids.
  • Amine-reactive succinimidyl ester
  • Blue fluorescent dye
  • Used for labeling proteins
  • Used for labeling nucleic acids