Learn More
Medchemexpress LLC 7-Hydroxycoumarin-3-carboxylic acid N-succinimidyl ester | 134471-24-2 | 50 MG
SDP
Catalog No. 5000410672
Encompass_Preferred
7-Hydroxycoumarin-3-carboxylic acid N-succinimidyl ester is the amine-reactive succinimidyl ester of 7-Hydroxycoumarin-3-carboxylic acid. It is a blue fluorescent dye used for labeling proteins and nucleic acids.
  • Amine-reactive succinimidyl ester
  • Blue fluorescent dye
  • For labeling proteins
  • For labeling nucleic acids
Catalog No. 50-004-10672 Supplier Medchemexpress LLC Supplier No. HYD011550MG
7-Hydroxycoumarin-3-carboxylic acid N-succinimidyl ester is the amine-reactive succinimidyl ester of 7-Hydroxycoumarin-3-carboxylic acid. It is a blue fluorescent dye used for labeling proteins and nucleic acids.
  • Amine-reactive succinimidyl ester
  • Blue fluorescent dye
  • For labeling proteins
  • For labeling nucleic acids