Learn More
Medchemexpress LLC 7-Hydroxycoumarin-3-carboxylic acid N-succinimidyl ester | 134471-24-2 | MFCD00037568 | ≥95.0% | 5 MG
SDP
Catalog No. 5000410673
Encompass_Preferred
7-Hydroxycoumarin-3-carboxylic acid N-succinimidyl ester is the amine-reactive succinimidyl ester of 7-Hydroxycoumarin-3-carboxylic acid. It is a blue fluorescent dye for labeling proteins and nucleic acids.
  • Fluorescent dye
  • Amine-reactive succinimidyl ester
  • Labels proteins and nucleic acids
Catalog No. 50-004-10673 Supplier Medchemexpress LLC Supplier No. HYD01155MG
7-Hydroxycoumarin-3-carboxylic acid N-succinimidyl ester is the amine-reactive succinimidyl ester of 7-Hydroxycoumarin-3-carboxylic acid. It is a blue fluorescent dye for labeling proteins and nucleic acids.
  • Fluorescent dye
  • Amine-reactive succinimidyl ester
  • Labels proteins and nucleic acids