Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Hydroxyflavanone, 99%

Catalog No. p-7027730
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2472003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2472003 Supplier Thermo Scientific Chemicals Supplier No. B2472003

CAS: 6515-36-2 | C15H12O3 | 240.258 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6515-36-2
Molecular Formula C15H12O3
Molecular Weight (g/mol) 240.258
MDL Number MFCD00017487
InChI Key SWAJPHCXKPCPQZ-UHFFFAOYSA-N
Synonym 7-hydroxyflavanone, 7-hydroxy-2-phenylchroman-4-one, chembl97542, 7-hydroxy-2-phenyl-4-chromanone, 7-hydroxy-2-phenyl-2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-7-hydroxy-2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2,3-dihydro-7-hydroxy-2-phenyl, 7-hydroxy-flavanone, d05zpo
PubChem CID 1890
ChEBI CHEBI:34483
IUPAC Name 7-hydroxy-2-phenyl-2,3-dihydrochromen-4-one
SMILES C1C(OC2=C(C1=O)C=CC(=C2)O)C3=CC=CC=C3

Specifications

Melting Point 187°C to 189°C
Quantity 1 g
Beilstein 87355
Formula Weight 240.25
Percent Purity 99%
Chemical Name or Material 7-Hydroxyflavanone
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.