Learn More
Medchemexpress LLC 7-Hydroxyflavone | 6665-86-7 | 1 ML
SDP
Catalog No. 5000419764
Encompass_Preferred
7-Hydroxyflavone is a flavonoid isolated from *Clerodendrum phlomidis* that exhibits anti-inflammatory activity. It protects renal cells from nicotine-associated cytotoxicity through the ERK/Nrf2/HO-1 pathway. This compound inhibits PKM2, COX-2, and 5-LOX, and is orally active.
  • Inhibits nicotine-induced cytotoxicity and reduces ROS production in renal cells
  • Inhibits EV71 3C protease, blocking viral replication
  • Decreases malondialdehyde and ROS levels, increases SOD and GSH levels in HepG2 cells
  • Inhibits inflammatory cytokines and reduces myocardial injury markers in rat models
  • Ameliorates Paclitaxel or Vinorelbine-induced neuropathic pain in mice models
  • Reduces fat accumulation, improves hepatic steatosis and oxidative stress in NAFLD models
Catalog No. 50-004-19764 Supplier Medchemexpress LLC Supplier No. HYN710810MM/1ML
7-Hydroxyflavone is a flavonoid isolated from *Clerodendrum phlomidis* that exhibits anti-inflammatory activity. It protects renal cells from nicotine-associated cytotoxicity through the ERK/Nrf2/HO-1 pathway. This compound inhibits PKM2, COX-2, and 5-LOX, and is orally active.
  • Inhibits nicotine-induced cytotoxicity and reduces ROS production in renal cells
  • Inhibits EV71 3C protease, blocking viral replication
  • Decreases malondialdehyde and ROS levels, increases SOD and GSH levels in HepG2 cells
  • Inhibits inflammatory cytokines and reduces myocardial injury markers in rat models
  • Ameliorates Paclitaxel or Vinorelbine-induced neuropathic pain in mice models
  • Reduces fat accumulation, improves hepatic steatosis and oxidative stress in NAFLD models