Learn More
Medchemexpress LLC 7-Hydroxyflavone | 6665-86-7 | 100 MG
SDP
Catalog No. 5000419447
Encompass_Preferred
7-Hydroxyflavone is a flavonoid isolated from *Clerodendrum phlomidis*, demonstrating anti-inflammatory activity. It protects renal cells from nicotine (NIC)-associated cytotoxicity through the ERK/Nrf2/HO-1 pathway, and inhibits PKM2, COX-2, and 5-LOX. This compound is orally active.
  • Isolated from *Clerodendrum phlomidis*
  • Protects renal cells from nicotine-associated cytotoxicity
  • Inhibits PKM2 with an IC50 of 2.12 μM
  • Inhibits COX-2 and 5-LOX with IC50s of 27 μg/mL and 33 μg/mL
  • Orally active
  • Ameliorates neuropathic pain in mice models
  • Improves high-fat diet-induced non-alcoholic fatty liver disease
  • Reduces malondialdehyde (MDA) and ROS levels in HepG2 cells
Catalog No. 50-004-19447 Supplier Medchemexpress LLC Supplier No. HYN7108100MG
7-Hydroxyflavone is a flavonoid isolated from *Clerodendrum phlomidis*, demonstrating anti-inflammatory activity. It protects renal cells from nicotine (NIC)-associated cytotoxicity through the ERK/Nrf2/HO-1 pathway, and inhibits PKM2, COX-2, and 5-LOX. This compound is orally active.
  • Isolated from *Clerodendrum phlomidis*
  • Protects renal cells from nicotine-associated cytotoxicity
  • Inhibits PKM2 with an IC50 of 2.12 μM
  • Inhibits COX-2 and 5-LOX with IC50s of 27 μg/mL and 33 μg/mL
  • Orally active
  • Ameliorates neuropathic pain in mice models
  • Improves high-fat diet-induced non-alcoholic fatty liver disease
  • Reduces malondialdehyde (MDA) and ROS levels in HepG2 cells