Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-Hydroxyflavone | 6665-86-7 | MFCD00006835 | 50 MG
SDP

Catalog No. 5000417281
Encompass_Preferred

7-Hydroxyflavone is a flavonoid isolated from *Clerodendrum phlomidis* that exhibits anti-inflammatory activity. It protects renal cells from nicotine-associated cytotoxicity via the ERK/Nrf2/HO-1 pathway. This orally active compound also inhibits PKM2, COX-2, and 5-LOX, and has demonstrated antioxidant activity and efficacy in various animal models.

  • Isolated from *Clerodendrum phlomidis*
  • Exhibits anti-inflammatory activity
  • Protects renal cells from nicotine-associated cytotoxicity
  • Inhibits PKM2, COX-2, and 5-LOX
  • Orally active
  • Reduces malondialdehyde (MDA) and reactive oxygen species (ROS) levels
  • Increases superoxide dismutase (SOD) and glutathione (GSH) levels
  • Ameliorates ischemia/reperfusion injury
  • Improves high-fat diet-induced non-alcoholic fatty liver disease and obesity
  • Purity of 99.89%
  • Appearance is a white to off-white solid

Catalog No. 50-004-17281 Supplier Medchemexpress LLC Supplier No. HYN710850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Hydroxyflavone is a flavonoid isolated from *Clerodendrum phlomidis* that exhibits anti-inflammatory activity. It protects renal cells from nicotine-associated cytotoxicity via the ERK/Nrf2/HO-1 pathway. This orally active compound also inhibits PKM2, COX-2, and 5-LOX, and has demonstrated antioxidant activity and efficacy in various animal models.

  • Isolated from *Clerodendrum phlomidis*
  • Exhibits anti-inflammatory activity
  • Protects renal cells from nicotine-associated cytotoxicity
  • Inhibits PKM2, COX-2, and 5-LOX
  • Orally active
  • Reduces malondialdehyde (MDA) and reactive oxygen species (ROS) levels
  • Increases superoxide dismutase (SOD) and glutathione (GSH) levels
  • Ameliorates ischemia/reperfusion injury
  • Improves high-fat diet-induced non-alcoholic fatty liver disease and obesity
  • Purity of 99.89%
  • Appearance is a white to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.