Learn More
Selleck Chemical LLC 7-Hydroxyflavone S5380-1g
SDP
Catalog No. 503109505
Encompass_Preferred
7-Hydroxyflavone is a potent inhibitor of CYP1A1 with a Ki value of 0 015 M and exhibits 6-fold greater selectivity for CYP1A1 over CYP1A2 It also has excellent antioxidant properties and great potential to be applied as medicines
Catalog No. 50-310-9505 Supplier Selleck Chemical LLC Supplier No. S53801G
7-Hydroxyflavone is a potent inhibitor of CYP1A1 with a Ki value of 0 015 M and exhibits 6-fold greater selectivity for CYP1A1 over CYP1A2 It also has excellent antioxidant properties and great potential to be applied as medicines