Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-hydroxymethyl-10,11-mdcpt-d5 | 428816-69-7 | 98.0% | 427.42 | C22H13D5N2O7 | 10 MG
SDP

Catalog No. 5000287788
Encompass_Preferred

Deuterium-labeled 7-hydroxymethyl-10,11-MDCPT-d5 is a stable isotope analog of a camptothecin-derived payload used for antibody-drug conjugate (ADC) research and as an internal standard or tracer in quantitative mass spectrometry. The compound is supplied as a high-purity solid intended for milligram-scale research applications and is typically stored at -20 °C, protected from light.

  • Deuterium-labeled tracer for accurate quantitation by mass spectrometry.
  • Applicable as an ADC payload in conjugation research.
  • High reported purity (98.0%).
  • Available in milligram-scale package sizes suitable for R&D.
  • Characterized by molecular weight 427.42 and formula C22H13D5N2O7.

Catalog No. 50-002-87788 Supplier Medchemexpress LLC Supplier No. HY156756S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deuterium-labeled 7-hydroxymethyl-10,11-MDCPT-d5 is a stable isotope analog of a camptothecin-derived payload used for antibody-drug conjugate (ADC) research and as an internal standard or tracer in quantitative mass spectrometry. The compound is supplied as a high-purity solid intended for milligram-scale research applications and is typically stored at -20 °C, protected from light.

  • Deuterium-labeled tracer for accurate quantitation by mass spectrometry.
  • Applicable as an ADC payload in conjugation research.
  • High reported purity (98.0%).
  • Available in milligram-scale package sizes suitable for R&D.
  • Characterized by molecular weight 427.42 and formula C22H13D5N2O7.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.