Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Hydroxyquinoline, 99%

Catalog No. AC418820050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC418820050 5 g Glass bottle
AC418820010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC418820050 Supplier Thermo Scientific Chemicals Supplier No. 418820050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 580-20-1 | C9H7NO | 145.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 580-20-1
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00016730
InChI Key QUUFTOGCVYWGNK-UHFFFAOYSA-N
Synonym 7-hydroxyquinoline, quinolin-7-ol, 7-quinolinol, unii-d2g6yuk7zj, 7-chinolinol, ccris 4332, d2g6yuk7zj, xcrppapdrubkrj-uhfffaoysa-n, quinolin-1-ium-7-olate, 7hydroxyquinoline
PubChem CID 11378
IUPAC Name 1H-quinolin-7-one
SMILES C1=CNC2=CC(=O)C=CC2=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 239°C to 244°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Formula Weight 145.16
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 7-Hydroxyquinoline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-457-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.