Learn More
Medchemexpress LLC 7-hydroxywarfarin-d5 | 94820-65-2 | 98.1% | 329.36 g/mol | C19H11D5O5 | 5 MG
SDP
Catalog No. 5000222959
Encompass_Preferred
7-Hydroxywarfarin-d5 is a deuterium-labeled isotopologue of 7-hydroxywarfarin provided for research applications as an analytical reference and internal standard. It is supplied as a white to off-white solid with specified purity and recommended storage conditions for long-term stability. Use is restricted to laboratory research and analytical workflows.
  • Deuterium-labeled internal standard for mass spectrometry and quantitative assays.
  • High purity suitable for analytical applications.
  • Available in small mg quantities for method development and validation.
  • Soluble in DMSO at high concentration; ultrasonic agitation recommended.
  • Store sealed at -20°C; stock solutions stable at -80°C for up to 6 months.
Catalog No. 50-002-22959 Supplier Medchemexpress LLC Supplier No. HY134938S5MG
7-Hydroxywarfarin-d5 is a deuterium-labeled isotopologue of 7-hydroxywarfarin provided for research applications as an analytical reference and internal standard. It is supplied as a white to off-white solid with specified purity and recommended storage conditions for long-term stability. Use is restricted to laboratory research and analytical workflows.
  • Deuterium-labeled internal standard for mass spectrometry and quantitative assays.
  • High purity suitable for analytical applications.
  • Available in small mg quantities for method development and validation.
  • Soluble in DMSO at high concentration; ultrasonic agitation recommended.
  • Store sealed at -20°C; stock solutions stable at -80°C for up to 6 months.