Learn More
Medchemexpress LLC 7-hydroxywarfarin-d5 | 94820-65-2 | 98.1% | C19H11D5O5 | 10MG
SDP
Catalog No. 5000259255
Encompass_Preferred
7-Hydroxywarfarin-d5 is a deuterium-labeled analogue of 7-hydroxywarfarin used as an internal standard and tracer for analytical and pharmacokinetic studies. It provides a mass-shifted reference compound to support accurate LC-MS/MS quantification of warfarin metabolites.
  • Deuterium-labeled internal standard for LC-MS/MS assays.
  • Mass-shifted analogue minimizes interference with analyte signals.
  • High purity (98.1%).
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Stable when stored sealed at -20°C; in solvent: -80°C up to 6 months.
  • Supplied as a white to off-white solid suitable for reference use.
Catalog No. 50-002-59255 Supplier Medchemexpress LLC Supplier No. HY134938S10MG
7-Hydroxywarfarin-d5 is a deuterium-labeled analogue of 7-hydroxywarfarin used as an internal standard and tracer for analytical and pharmacokinetic studies. It provides a mass-shifted reference compound to support accurate LC-MS/MS quantification of warfarin metabolites.
  • Deuterium-labeled internal standard for LC-MS/MS assays.
  • Mass-shifted analogue minimizes interference with analyte signals.
  • High purity (98.1%).
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Stable when stored sealed at -20°C; in solvent: -80°C up to 6 months.
  • Supplied as a white to off-white solid suitable for reference use.