Learn More
Medchemexpress LLC 7-iodo-2',3'-dideoxy-7-deaza-guanosine | 114748-67-3 | 376.15 g/mol | C11H13IN4O3 | 10 MG
SDP
Catalog No. 5000210075
Encompass_Preferred
7-iodo-2',3'-dideoxy-7-deaza-guanosine is an iodinated dideoxynucleoside used as a chain-terminating reagent in DNA synthesis and sequencing applications. It is supplied for research use and should be handled and stored according to recommended conditions to preserve stability.
  • Iodinated dideoxynucleoside analog suitable for chain termination.
  • Used in DNA synthesis and sequencing reactions.
  • Available in small milligram pack sizes for research applications.
  • Molecular formula C11H13IN4O3 and molecular weight 376.15 g/mol.
  • Store protected from light at 4°C; in solvent store at -80°C for long-term.
Catalog No. 50-002-10075 Supplier Medchemexpress LLC Supplier No. HYW04847910MG
7-iodo-2',3'-dideoxy-7-deaza-guanosine is an iodinated dideoxynucleoside used as a chain-terminating reagent in DNA synthesis and sequencing applications. It is supplied for research use and should be handled and stored according to recommended conditions to preserve stability.
  • Iodinated dideoxynucleoside analog suitable for chain termination.
  • Used in DNA synthesis and sequencing reactions.
  • Available in small milligram pack sizes for research applications.
  • Molecular formula C11H13IN4O3 and molecular weight 376.15 g/mol.
  • Store protected from light at 4°C; in solvent store at -80°C for long-term.