Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-iodo-2',3'-dideoxy-7-deaza-guanosine | 114748-67-3 | 376.15 g/mol | C11H13IN4O3 | 5 MG
SDP

Catalog No. 5000209879
Encompass_Preferred

7-Iodo-2',3'-dideoxy-7-deaza-guanosine is a modified dideoxynucleoside used as a chain-terminating nucleotide analogue in DNA synthesis and sequencing reactions.

  • Modified dideoxynucleoside for chain termination in DNA sequencing
  • CAS number 114748-67-3
  • Molecular weight 376.15 g/mol
  • Molecular formula C11H13IN4O3
  • Store at 4°C; in solvent: -80°C (6 months), -20°C (1 month); protect from light
  • Available in milligram pack sizes, including 5 MG

Catalog No. 50-002-09879 Supplier Medchemexpress LLC Supplier No. HYW0484795MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Iodo-2',3'-dideoxy-7-deaza-guanosine is a modified dideoxynucleoside used as a chain-terminating nucleotide analogue in DNA synthesis and sequencing reactions.

  • Modified dideoxynucleoside for chain termination in DNA sequencing
  • CAS number 114748-67-3
  • Molecular weight 376.15 g/mol
  • Molecular formula C11H13IN4O3
  • Store at 4°C; in solvent: -80°C (6 months), -20°C (1 month); protect from light
  • Available in milligram pack sizes, including 5 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.