Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-iodo-2',3'-dideoxy-7-deaza-guanosine | 114748-67-3 | 99.2% | 376.15 g/mol | C11H13IN4O3 | 25 MG
SDP

Catalog No. 5000209709
Encompass_Preferred

7-iodo-2',3'-dideoxy-7-deaza-guanosine is a dideoxynucleoside analog used as a chain-terminating nucleotide in DNA synthesis and sequencing reactions. It is supplied as a solid and as a solution in DMSO and is used for chain-termination studies and sequencing reaction termination.

  • Chain-terminating dideoxynucleoside for DNA synthesis and sequencing.
  • High purity suitable for molecular biology and analytical applications.
  • Available in multiple pack sizes and as a 10 mM solution in DMSO for convenience.
  • Molecular weight 376.15 g/mol and formula C11H13IN4O3.

Catalog No. 50-002-09709 Supplier Medchemexpress LLC Supplier No. 25MGHYW048479
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-iodo-2',3'-dideoxy-7-deaza-guanosine is a dideoxynucleoside analog used as a chain-terminating nucleotide in DNA synthesis and sequencing reactions. It is supplied as a solid and as a solution in DMSO and is used for chain-termination studies and sequencing reaction termination.

  • Chain-terminating dideoxynucleoside for DNA synthesis and sequencing.
  • High purity suitable for molecular biology and analytical applications.
  • Available in multiple pack sizes and as a 10 mM solution in DMSO for convenience.
  • Molecular weight 376.15 g/mol and formula C11H13IN4O3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.