Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-iodo-2',3'-dideoxy-7-deaza-guanosine | 114748-67-3 | 99.2% | 376.15 g/mol | C11H13IN4O3 | 50 MG
SDP

Catalog No. 5000210882
Encompass_Preferred

7-iodo-2',3'-dideoxy-7-deaza-guanosine is an iodinated dideoxynucleoside used as a chain-terminating nucleotide in DNA synthesis and sequencing reactions. It is supplied as a solid and is also available as a 10 mM solution in DMSO; analytical purity is listed at 99.17%.

  • used as a chain terminator in DNA synthesis and sequencing
  • available as a solid and as a 10 mM solution in DMSO
  • high analytical purity suitable for research applications
  • molecular weight 376.15 g/mol and formula C11H13IN4O3
  • CAS number 114748-67-3

Catalog No. 50-002-10882 Supplier Medchemexpress LLC Supplier No. HYW04847950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-iodo-2',3'-dideoxy-7-deaza-guanosine is an iodinated dideoxynucleoside used as a chain-terminating nucleotide in DNA synthesis and sequencing reactions. It is supplied as a solid and is also available as a 10 mM solution in DMSO; analytical purity is listed at 99.17%.

  • used as a chain terminator in DNA synthesis and sequencing
  • available as a solid and as a 10 mM solution in DMSO
  • high analytical purity suitable for research applications
  • molecular weight 376.15 g/mol and formula C11H13IN4O3
  • CAS number 114748-67-3

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.